Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MATN4
Entrez Gene ID
8785
Gene Name
matrilin 4
Gene Aliases
-
Chromosome Location
Chr.20: 45293446 - 45308566 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003833.4 NP_003824.2 4 - 5 885 181
NM_030590.3 NP_085080.1 3 - 4 58
XM_005260597.1 XP_005260654.1 3 - 4 1066 181
XM_017028113.1 XP_016883602.1 4 - 5 885 181
XM_017028114.1 XP_016883603.1 3 - 4 58
GenBank mRNA AJ007581.1 - 2 - 3 649 181
AK074495.1 - 1 - 2 58
AK074593.1 - 4 - 6 181
AK074595.1 - 3 - 4 58
AK074597.1 - 3 - 4 58
BC131763.1 - 2 - 3 653 181
BC141811.1 - 3 - 4 58
BC143625.1 - 2 - 3 612 181
BC151219.1 - 3 - 4 780 181

View Full Product Details