Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NFS1
Entrez Gene ID
9054
Gene Name
NFS1 cysteine desulfurase
Gene Aliases
HUSSY-08, IscS, NIFS
Chromosome Location
Chr.20: 35668688 - 35699365 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198989.1 NP_001185918.1 11 - 12 1237 85
NM_021100.4 NP_066923.3 12 - 13 1390 85
NR_037570.2 13 - 14 1512 85
GenBank mRNA AF097025.2 - 12 - 13 1376 85
AJ010952.1 - 8 - 9 898 85
AK001265.1 - 12 - 13 1375 85
AK001470.1 - 12 - 13 1386 85
AK056242.1 - 12 - 13 1661 85
AK091049.1 - 4 - 5 1932 85
AK223239.1 - 12 - 13 1376 85
AK297969.1 - 13 - 14 1499 85
AK301969.1 - 9 - 10 818 85
AK302023.1 - 11 - 12 1224 85
AK312293.1 - 12 - 13 1377 85
BC065560.1 - 12 - 13 1362 85

View Full Product Details