The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198863.1 | NP_001185792.1 | 15 - 16 | 1618 | 72 |
NM_003915.5 | NP_003906.2 | 15 - 16 | 1848 | 72 | |
NM_152925.2 | NP_690902.1 | 15 - 16 | 1663 | 72 | |
NM_152926.2 | NP_690903.1 | 15 - 16 | 1621 | 72 | |
NM_152927.2 | NP_690904.1 | 16 - 17 | 1748 | 72 | |
NM_152928.2 | NP_690905.1 | 16 - 17 | 1716 | 72 | |
NR_037188.1 | 17 - 18 | 1840 | 72 | ||
GenBank mRNA | AB209827.1 | - | 15 - 16 | 1594 | 72 |
AF314092.1 | - | 4 - 5 | 474 | 72 | |
AI075146.1 | - | 1 - 2 | 72 | ||
BC001142.1 | - | 17 - 18 | 1789 | 72 | |
BC021010.1 | - | 15 - 16 | 1567 | 72 | |
BG231745.1 | - | 1 - 2 | 72 | ||
BQ433414.1 | - | - | 354 | 72 | |
CR456790.1 | - | 14 - 15 | 1466 | 72 | |
U83246.1 | - | 15 - 16 | 1622 | 72 |