The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033118.3 | NP_149109.1 | 5 - 6 | 978 | 78 |
GenBank mRNA | AF325549.1 | - | 4 - 5 | 876 | 78 |
AJ272502.1 | - | 5 - 6 | 984 | 78 | |
BC069627.1 | - | 4 - 5 | 876 | 78 | |
BC092413.1 | - | 5 - 6 | 978 | 78 | |
BC127622.1 | - | 4 - 5 | 876 | 78 |