The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002646.3 | NP_002637.3 | 17 - 18 | 3152 | 83 |
XM_005245257.2 | XP_005245314.1 | 16 - 17 | 3217 | 83 | |
XM_005245258.3 | XP_005245315.1 | 17 - 18 | 3136 | 83 | |
XM_011509630.2 | XP_011507932.1 | 18 - 19 | 4011 | 83 | |
XM_011509631.2 | XP_011507933.1 | 17 - 18 | 3328 | 83 | |
XM_017001473.1 | XP_016856962.1 | 17 - 18 | 4147 | 83 | |
XM_017001474.1 | XP_016856963.1 | 17 - 18 | 3350 | 83 | |
XM_017001475.1 | XP_016856964.1 | 15 - 16 | 2098 | 83 | |
GenBank mRNA | AB210006.1 | - | 15 - 16 | 2706 | 83 |
BC132876.1 | - | 17 - 18 | 3124 | 83 | |
BC136645.1 | - | 17 - 18 | 3124 | 83 | |
BC144342.1 | - | 18 - 19 | 3040 | 83 | |
Y11312.1 | - | 17 - 18 | 3188 | 83 | |
Y13892.1 | - | 15 - 16 | 2689 | 83 |