The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002646.3 | NP_002637.3 | 27 - 28 | 4431 | 116 |
XM_005245257.2 | XP_005245314.1 | 26 - 27 | 4496 | 116 | |
XM_005245258.3 | XP_005245315.1 | 27 - 28 | 4415 | 116 | |
XM_011509630.2 | XP_011507932.1 | 28 - 29 | 5290 | 116 | |
XM_011509631.2 | XP_011507933.1 | 27 - 28 | 4607 | 116 | |
XM_017001473.1 | XP_016856962.1 | 27 - 28 | 5426 | 116 | |
XM_017001474.1 | XP_016856963.1 | 27 - 28 | 4629 | 116 | |
XM_017001475.1 | XP_016856964.1 | 25 - 26 | 3377 | 116 | |
GenBank mRNA | AB210006.1 | - | 25 - 26 | 3985 | 116 |
BC132876.1 | - | 27 - 28 | 4403 | 116 | |
BC136645.1 | - | 27 - 28 | 4403 | 116 | |
BC144342.1 | - | 28 - 29 | 4319 | 116 | |
Y11312.1 | - | 27 - 28 | 4467 | 116 | |
Y13892.1 | - | 25 - 26 | 3968 | 116 |