Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PIK3C2B
Entrez Gene ID
5287
Gene Name
phosphatidylinositol-4-phosphate 3-kinase catalytic subunit type 2 beta
Gene Aliases
C2-PI3K
Chromosome Location
Chr.1: 204422630 - 204494815 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002646.3 NP_002637.3 33 - 34 5197 113
XM_005245257.2 XP_005245314.1 32 - 33 5262 113
XM_005245258.3 XP_005245315.1 33 - 34 5181 113
XM_011509630.2 XP_011507932.1 34 - 35 6056 113
XM_011509631.2 XP_011507933.1 33 - 34 5373 113
XM_017001473.1 XP_016856962.1 33 - 34 6192 113
XM_017001474.1 XP_016856963.1 33 - 34 5395 113
XM_017001475.1 XP_016856964.1 31 - 32 4143 113
GenBank mRNA AB210006.1 - 31 - 32 4751 113
BC132876.1 - 33 - 34 5169 113
BC136645.1 - 33 - 34 5169 113
BC144342.1 - 34 - 35 5085 113
Y11312.1 - 33 - 34 5233 113
Y13892.1 - 31 - 32 4734 113

View Full Product Details