The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007144.2 | NP_009075.1 | 7 - 8 | 668 | 81 |
XM_005257640.2 | XP_005257697.1 | 8 - 9 | 1127 | 81 | |
XM_005257641.4 | XP_005257698.1 | 6 - 7 | 1349 | 81 | |
XM_005257642.3 | XP_005257699.1 | 6 - 7 | 1746 | 81 | |
XM_017025016.1 | XP_016880505.1 | 8 - 9 | 1687 | 81 | |
GenBank mRNA | BC004858.2 | - | 5 - 6 | 439 | 81 |
BC024255.1 | - | 7 - 8 | 664 | 81 | |
BX647429.1 | - | 8 - 9 | 1127 | 81 | |
D13969.1 | - | 6 - 7 | 81 |