Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FCN3
Entrez Gene ID
8547
Gene Name
ficolin 3
Gene Aliases
FCNH, HAKA1
Chromosome Location
Chr.1: 27369110 - 27374824 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK075140.1 - 2 - 3 193 115
AK309540.1 - 2 - 3 189 148
AY358081.1 - 2 - 3 182 115
AY358283.1 - 2 - 3 219 115
BC020731.1 - 2 - 3 193 115
BG546674.1 - 2 - 3 222 148
CR456808.1 - 2 - 3 187 148

View Full Product Details