Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TECTA
Entrez Gene ID
7007
Gene Name
tectorin alpha
Gene Aliases
DFNA12, DFNA8, DFNB21
Chromosome Location
Chr.11: 121102666 - 121190806 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005422.2 NP_005413.2 16 - 17 5378 71

View Full Product Details