The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001257191.2 | NP_001244120.1 | 7 - 8 | 1096 | 148 |
NM_004397.5 | NP_004388.2 | 7 - 8 | 1168 | 148 | |
XM_005271417.3 | XP_005271474.1 | 7 - 8 | 1113 | 148 | |
XM_011542644.1 | XP_011540946.1 | 7 - 8 | 1168 | 148 | |
XM_011542645.1 | XP_011540947.1 | 6 - 7 | 1021 | 148 | |
XM_017017251.1 | XP_016872740.1 | 7 - 8 | 1113 | 148 | |
GenBank mRNA | AK313244.1 | - | 7 - 8 | 1096 | 148 |
BC065007.1 | - | 6 - 7 | 1027 | 148 | |
D17532.1 | - | 7 - 8 | 1041 | 148 | |
Z11685.1 | - | 7 - 8 | 1068 | 148 |