Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SPEG
- Entrez Gene ID
-
10290
- Gene Name
- SPEG complex locus
- Gene Aliases
-
APEG-1, APEG1, BPEG, CNM5, SPEGalpha, SPEGbeta
- Chromosome Location
-
Chr.2: 219434215 - 219493632 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_005876.4 |
NP_005867.3 |
1 - 2 |
393 |
73 |
View Full Product Details