The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144830.2 | NP_001138302.1 | 5 - 6 | 1445 | 116 |
XM_006711245.3 | XP_006711308.1 | 6 - 7 | 1215 | 116 | |
GenBank mRNA | BC035687.1 | - | 5 - 6 | 1118 | 116 |