The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033225.5 | NP_150094.5 | 31 - 32 | 5376 | 66 |
XM_011534752.2 | XP_011533054.1 | 31 - 32 | 5605 | 66 | |
XM_011534753.2 | XP_011533055.1 | 14 - 15 | 2483 | 66 | |
XM_011534754.1 | XP_011533056.1 | 7 - 8 | 1219 | 66 | |
XM_017013731.1 | XP_016869220.1 | 31 - 32 | 5605 | 66 | |
GenBank mRNA | AB067477.1 | - | 12 - 13 | 2094 | 66 |
AB209502.1 | - | 19 - 20 | 3428 | 66 | |
AF333704.2 | - | 32 - 33 | 5379 | 66 | |
AK126936.1 | - | 7 - 8 | 1219 | 66 | |
AY017307.1 | - | 32 - 33 | 5274 | 66 | |
AY358174.1 | - | 15 - 16 | 2519 | 66 |