Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CSMD1
Entrez Gene ID
64478
Gene Name
CUB and Sushi multiple domains 1
Gene Aliases
PPP1R24
Chromosome Location
Chr.8: 2935353 - 4995035 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033225.5 NP_150094.5 40 - 41 6422 114
XM_011534752.2 XP_011533054.1 40 - 41 6651 114
XM_011534753.2 XP_011533055.1 23 - 24 3529 114
XM_011534754.1 XP_011533056.1 16 - 17 2265 114
XM_017013731.1 XP_016869220.1 40 - 41 6651 114
GenBank mRNA AB209502.1 - 28 - 29 4474 114
AF333704.2 - 41 - 42 6428 114
AK126936.1 - 16 - 17 2265 114
AY017307.1 - 41 - 42 6323 114

View Full Product Details