The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_033169.1 | 4 - 5 | 743 | 144 | |
XM_011515852.2 | XP_011514154.1 | 2 - 3 | 317 | 144 | |
GenBank mRNA | AK093091.1 | - | 4 - 5 | 696 | 144 |
AK315667.1 | - | 4 - 5 | 703 | 144 |