Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VPS33B
Entrez Gene ID
26276
Gene Name
VPS33B, late endosome and lysosome associated
Gene Aliases
-
Chromosome Location
Chr.15: 90998416 - 91022621 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289148.1 NP_001276077.1 2 - 3 535 80
NM_018668.4 NP_061138.3 3 - 4 616 80
XM_011521448.1 XP_011519750.1 3 - 4 638 80
XM_017022075.1 XP_016877564.1 3 - 4 455 80
GenBank mRNA AF201694.1 - 3 - 4 547 80
AF308803.1 - 3 - 4 580 80
AK027754.1 - 3 - 4 578 80
AK293688.1 - 2 - 3 510 80
AK300608.1 - 4 - 5 281 80
AK308480.1 - 3 - 4 638 80
AK315721.1 - 3 - 4 570 80
BC016445.1 - 3 - 4 451 80
KU178590.1 - 3 - 4 244 80

View Full Product Details