The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289148.1 | NP_001276077.1 | 8 - 9 | 996 | 84 |
NM_001289149.1 | NP_001276078.1 | 8 - 9 | 1015 | 84 | |
NM_018668.4 | NP_061138.3 | 9 - 10 | 1077 | 84 | |
XM_005254887.1 | XP_005254944.1 | 7 - 8 | 909 | 84 | |
XM_011521448.1 | XP_011519750.1 | 9 - 10 | 1099 | 84 | |
XM_011521449.2 | XP_011519751.1 | 5 - 6 | 396 | 84 | |
XM_017022075.1 | XP_016877564.1 | 9 - 10 | 932 | 84 | |
XM_017022076.1 | XP_016877565.1 | 7 - 8 | 925 | 84 | |
GenBank mRNA | AF201694.1 | - | 9 - 10 | 1008 | 84 |
AF308803.1 | - | 9 - 10 | 1041 | 84 | |
AK027754.1 | - | 9 - 10 | 1039 | 84 | |
AK074863.1 | - | 8 - 9 | 908 | 84 | |
AK293688.1 | - | 8 - 9 | 971 | 84 | |
AK300608.1 | - | 10 - 11 | 742 | 84 | |
AK315721.1 | - | 9 - 10 | 1028 | 84 | |
AL357472.1 | - | 6 - 7 | 439 | 84 | |
BC016445.1 | - | 9 - 10 | 912 | 84 | |
KU178590.1 | - | 9 - 10 | 705 | 84 | |
KU178591.1 | - | 7 - 8 | 578 | 84 |