Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ERGIC3
Entrez Gene ID
51614
Gene Name
ERGIC and golgi 3
Gene Aliases
C20orf47, CGI-54, Erv46, NY-BR-84, PRO0989, SDBCAG84, dJ477O4.2
Chromosome Location
Chr.20: 35542029 - 35557634 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015966.2 NP_057050.1 3 - 4 309 117
NM_198398.1 NP_938408.1 3 - 4 309 117
GenBank mRNA AF077030.1 - 3 - 4 267 117
AF116614.1 - 3 - 4 416 117
AF151812.1 - 3 - 4 267 117
AF308298.1 - 3 - 4 269 117
AK074550.1 - 3 - 4 257 117
AK096484.1 - 3 - 4 257 117
AK300916.1 - 3 - 4 258 117
BC009765.1 - 3 - 4 263 117
BC014014.2 - 3 - 4 262 117
BU528333.1 - 3 - 4 286 117
CA489637.1 - 3 - 4 309 117
EF206690.1 - 3 - 4 267 117

View Full Product Details