Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ERGIC3
Entrez Gene ID
51614
Gene Name
ERGIC and golgi 3
Gene Aliases
C20orf47, CGI-54, Erv46, NY-BR-84, PRO0989, SDBCAG84, dJ477O4.2
Chromosome Location
Chr.20: 35542029 - 35557634 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015966.2 NP_057050.1 5 - 6 530 68
NM_198398.1 NP_938408.1 5 - 6 530 68
GenBank mRNA AF077030.1 - 5 - 6 488 68
AF116614.1 - 5 - 6 637 68
AF151812.1 - 5 - 6 488 68
AF308298.1 - 5 - 6 490 68
AK074550.1 - 5 - 6 478 68
AK300916.1 - 5 - 6 479 68
BC009765.1 - 5 - 6 484 68
BC014014.2 - 5 - 6 483 68
BU528333.1 - 5 - 6 507 68
CA489637.1 - 5 - 6 530 68
EF206690.1 - 5 - 6 488 68

View Full Product Details