The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021819.2 | NP_068591.2 | 5 - 6 | 605 | 96 |
GenBank mRNA | AF303398.1 | - | 5 - 6 | 641 | 96 |
AK291819.1 | - | 5 - 6 | 610 | 96 | |
AK294807.1 | - | 5 - 6 | 446 | 96 | |
AK300517.1 | - | 6 - 7 | 560 | 96 | |
AK309976.1 | - | 6 - 7 | 537 | 96 | |
AY358724.1 | - | 5 - 6 | 605 | 96 |