Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CABIN1
Entrez Gene ID
23523
Gene Name
calcineurin binding protein 1
Gene Aliases
CAIN, KB-318B8.7, PPP3IN
Chromosome Location
Chr.22: 24011192 - 24178628 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199281.1 NP_001186210.1 25 - 26 4378 82
NM_001201429.1 NP_001188358.1 24 - 25 3922 82
NM_012295.3 NP_036427.1 25 - 26 4072 82
XM_005261415.2 XP_005261472.1 24 - 25 4019 82
XM_005261416.2 XP_005261473.1 24 - 25 4019 82
XM_005261417.2 XP_005261474.1 24 - 25 4019 82
XM_005261419.2 XP_005261476.1 24 - 25 4060 82
XM_006724179.1 XP_006724242.1 24 - 25 4019 82
XM_006724180.1 XP_006724243.1 24 - 25 4019 82
XM_006724181.1 XP_006724244.1 24 - 25 4019 82
XM_011530026.1 XP_011528328.1 23 - 24 3824 82
XM_011530029.2 XP_011528331.1 24 - 25 4019 82
XM_011530030.2 XP_011528332.1 24 - 25 4019 82
XM_017028680.1 XP_016884169.1 24 - 25 4019 82
XM_017028681.1 XP_016884170.1 23 - 24 3869 82
XM_017028682.1 XP_016884171.1 24 - 25 3940 82
XM_017028683.1 XP_016884172.1 23 - 24 3869 82
XM_017028684.1 XP_016884173.1 9 - 10 1557 82
GenBank mRNA AB002328.2 - 25 - 26 4295 82
AF072441.1 - 25 - 26 4073 82
BC054497.1 - 24 - 25 3889 82

View Full Product Details