The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145938.1 | NP_001139410.1 | 8 - 9 | 1187 | 128 |
NM_002421.3 | NP_002412.1 | 8 - 9 | 1339 | 128 | |
GenBank mRNA | AK024818.1 | - | 8 - 9 | 128 | |
AK130687.1 | - | 8 - 9 | 1262 | 128 | |
AK223034.1 | - | 7 - 8 | 965 | 128 | |
AK223035.1 | - | 8 - 9 | 1220 | 128 | |
AK223036.1 | - | 6 - 7 | 803 | 128 | |
AK223056.1 | - | 7 - 8 | 1005 | 128 | |
AK223281.1 | - | 8 - 9 | 1262 | 128 | |
AK225495.1 | - | 7 - 8 | 1030 | 128 | |
AK297723.1 | - | 8 - 9 | 1110 | 128 | |
BC013118.1 | - | 4 - 5 | 487 | 128 | |
BC013875.2 | - | 8 - 9 | 1238 | 128 | |
BT006874.1 | - | 8 - 9 | 1196 | 128 | |
BT020147.1 | - | 8 - 9 | 1196 | 128 | |
X05231.1 | - | 8 - 9 | 1262 | 128 | |
X54925.1 | - | 8 - 9 | 1264 | 128 |