The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005740.2 | NP_005731.1 | 3 - 4 | 383 | 96 |
GenBank mRNA | AK223349.1 | - | 3 - 4 | 370 | 96 |
AK290566.1 | - | 3 - 4 | 336 | 96 | |
AL035366.1 | - | 3 - 4 | 376 | 96 | |
AY453399.1 | - | - | 174 | 96 | |
BC002968.1 | - | 3 - 4 | 350 | 96 | |
BT009847.1 | - | 2 - 3 | 155 | 96 | |
BX473356.1 | - | 3 - 4 | 388 | 96 | |
CR456487.1 | - | 2 - 3 | 240 | 96 | |
CR542177.1 | - | 2 - 3 | 155 | 96 |