Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDH4
Entrez Gene ID
1002
Gene Name
cadherin 4
Gene Aliases
CAD4, R-CAD, RCAD
Chromosome Location
Chr.20: 61252426 - 61940617 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252338.2 NP_001239267.1 12 - 13 2145 67
NM_001252339.2 NP_001239268.1 12 - 13 2302 67
NM_001794.4 NP_001785.2 13 - 14 2330 67
GenBank mRNA AK091496.1 - 13 - 14 2526 67
AK124724.1 - 12 - 13 2285 67
AK308345.1 - 12 - 13 2144 67
AK315192.1 - 13 - 14 2284 67
BC101651.1 - 13 - 14 2330 67
BC112150.1 - 13 - 14 2330 67
L34059.1 - 13 - 14 2256 67

View Full Product Details