The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252338.2 | NP_001239267.1 | 14 - 15 | 2449 | 84 |
NM_001252339.2 | NP_001239268.1 | 14 - 15 | 2606 | 84 | |
NM_001794.4 | NP_001785.2 | 15 - 16 | 2634 | 84 | |
GenBank mRNA | AK091496.1 | - | 15 - 16 | 2830 | 84 |
AK124724.1 | - | 14 - 15 | 2589 | 84 | |
AK308345.1 | - | 14 - 15 | 2448 | 84 | |
AK315192.1 | - | 15 - 16 | 2588 | 84 | |
BC101651.1 | - | 15 - 16 | 2634 | 84 | |
BC112150.1 | - | 15 - 16 | 2634 | 84 | |
L34059.1 | - | 15 - 16 | 2560 | 84 |