The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252338.2 | NP_001239267.1 | 7 - 8 | 1096 | 65 |
NM_001252339.2 | NP_001239268.1 | 7 - 8 | 1253 | 65 | |
NM_001794.4 | NP_001785.2 | 8 - 9 | 1281 | 65 | |
GenBank mRNA | AK091496.1 | - | 8 - 9 | 1477 | 65 |
AK124724.1 | - | 7 - 8 | 1236 | 65 | |
AK308345.1 | - | 7 - 8 | 1095 | 65 | |
AK315192.1 | - | 8 - 9 | 1235 | 65 | |
BC101651.1 | - | 8 - 9 | 1281 | 65 | |
BC112150.1 | - | 8 - 9 | 1281 | 65 |