Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MORC3
Entrez Gene ID
23515
Gene Name
MORC family CW-type zinc finger 3
Gene Aliases
NXP2, ZCW5, ZCWCC3
Chromosome Location
Chr.21: 36320189 - 36376646 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320445.1 NP_001307374.1 4 - 5 608 114
NM_001320446.1 NP_001307375.1 6 - 7 767 114
NM_015358.2 NP_056173.1 5 - 6 681 114
XM_011529518.2 XP_011527820.1 5 - 6 901 114
XM_017028311.1 XP_016883800.1 5 - 6 703 114
XM_017028312.1 XP_016883801.1 5 - 6 624 114
GenBank mRNA AK292957.1 - 5 - 6 632 114
AK295133.1 - 4 - 5 600 114
BC094779.1 - 5 - 6 638 114
BC132731.1 - 5 - 6 635 114
BC144116.1 - 6 - 7 721 114
D50926.2 - 5 - 6 640 114

View Full Product Details