The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100397.1 | NP_001093867.1 | 16 - 17 | 1848 | 86 |
NM_001282899.1 | NP_001269828.1 | 16 - 17 | 1719 | 86 | |
NM_001282900.1 | NP_001269829.1 | 15 - 16 | 1665 | 86 | |
NM_001282901.1 | NP_001269830.1 | 13 - 14 | 1453 | 86 | |
NM_007023.3 | NP_008954.2 | 19 - 20 | 2089 | 86 | |
XM_005246246.2 | XP_005246303.1 | 18 - 19 | 1902 | 86 | |
XM_006712205.3 | XP_006712268.1 | 18 - 19 | 1907 | 86 | |
XM_011510505.2 | XP_011508807.1 | 18 - 19 | 1936 | 86 | |
XM_017003196.1 | XP_016858685.1 | 19 - 20 | 1956 | 86 | |
XM_017003197.1 | XP_016858686.1 | 15 - 16 | 1605 | 86 | |
GenBank mRNA | AB027471.1 | - | 19 - 20 | 1959 | 86 |
AB075513.1 | - | 3 - 4 | 304 | 86 | |
AB209681.1 | - | 19 - 20 | 4143 | 86 | |
AK294433.1 | - | 13 - 14 | 1377 | 86 | |
AK294445.1 | - | 16 - 17 | 1568 | 86 | |
AK294914.1 | - | 13 - 14 | 1406 | 86 | |
AK295002.1 | - | 15 - 16 | 1578 | 86 | |
AK296340.1 | - | 15 - 16 | 1660 | 86 | |
AK304278.1 | - | 16 - 17 | 1608 | 86 | |
AK313084.1 | - | 19 - 20 | 1958 | 86 | |
AK316014.1 | - | 16 - 17 | 1683 | 86 | |
AK316072.1 | - | 16 - 17 | 1615 | 86 | |
BC024004.2 | - | 19 - 20 | 1928 | 86 | |
KJ534924.1 | - | 19 - 20 | 1866 | 86 | |
KJ534925.1 | - | 18 - 19 | 1848 | 86 | |
KJ534978.1 | - | 19 - 20 | 1902 | 86 | |
U78516.1 | - | 19 - 20 | 2010 | 86 |