The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100397.1 | NP_001093867.1 | 27 - 28 | 2850 | 69 |
NM_001282899.1 | NP_001269828.1 | 27 - 28 | 2721 | 69 | |
NM_001282900.1 | NP_001269829.1 | 26 - 27 | 2667 | 69 | |
NM_001282901.1 | NP_001269830.1 | 24 - 25 | 2455 | 69 | |
NM_007023.3 | NP_008954.2 | 30 - 31 | 3091 | 69 | |
XM_005246246.2 | XP_005246303.1 | 29 - 30 | 2904 | 69 | |
XM_006712205.3 | XP_006712268.1 | 29 - 30 | 2909 | 69 | |
XM_011510505.2 | XP_011508807.1 | 29 - 30 | 2938 | 69 | |
XM_017003197.1 | XP_016858686.1 | 26 - 27 | 2607 | 69 | |
GenBank mRNA | AB027471.1 | - | 30 - 31 | 2961 | 69 |
AB075513.1 | - | 14 - 15 | 1306 | 69 | |
AB209681.1 | - | 30 - 31 | 5145 | 69 | |
AK294433.1 | - | 24 - 25 | 2379 | 69 | |
AK294445.1 | - | 27 - 28 | 2570 | 69 | |
AK294914.1 | - | 24 - 25 | 2408 | 69 | |
AK296340.1 | - | 26 - 27 | 2662 | 69 | |
AK304278.1 | - | 27 - 28 | 2610 | 69 | |
AK313084.1 | - | 30 - 31 | 2960 | 69 | |
AK316014.1 | - | 27 - 28 | 2685 | 69 | |
AK316072.1 | - | 27 - 28 | 2617 | 69 | |
BC024004.2 | - | 30 - 31 | 2930 | 69 | |
BC040183.2 | - | 27 - 28 | 2851 | 69 | |
KJ534924.1 | - | 30 - 31 | 2868 | 69 | |
KJ534925.1 | - | 29 - 30 | 2850 | 69 | |
KJ534978.1 | - | 30 - 31 | 2904 | 69 | |
U78516.1 | - | 30 - 31 | 3012 | 69 |