Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYP39A1
Entrez Gene ID
51302
Gene Name
cytochrome P450 family 39 subfamily A member 1
Gene Aliases
-
Chromosome Location
Chr.6: 46549580 - 46652849 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278738.1 NP_001265667.1 1 - 2 432 84
NM_001278739.1 NP_001265668.1 1 - 2 432 84
NM_016593.4 NP_057677.2 1 - 2 432 84
XM_005249171.3 XP_005249228.1 1 - 2 407 84
XM_011514658.1 XP_011512960.1 1 - 2 407 84
XM_017010921.1 XP_016866410.1 1 - 2 407 84
XM_017010922.1 XP_016866411.1 1 - 2 451 84
XM_017010923.1 XP_016866412.1 1 - 2 407 84
XM_017010924.1 XP_016866413.1 1 - 2 407 84
XM_017010925.1 XP_016866414.1 1 - 2 407 84
GenBank mRNA AF237982.1 - 1 - 2 228 84
AK292263.1 - 1 - 2 364 84
AK301604.1 - 1 - 2 310 84
AK307986.1 - 1 - 2 95 84
BC010358.1 - 1 - 2 392 84
DC336695.1 - 1 - 2 432 84

View Full Product Details