The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000601.5 | NP_000592.3 | 8 - 9 | 1267 | 72 |
NM_001010932.2 | NP_001010932.1 | 8 - 9 | 1252 | 72 | |
XM_006715956.2 | XP_006716019.1 | 9 - 10 | 1230 | 72 | |
XM_011516115.2 | XP_011514417.1 | 9 - 10 | 1215 | 72 | |
GenBank mRNA | AK291674.1 | - | 8 - 9 | 1202 | 72 |
AK313694.1 | - | 8 - 9 | 1102 | 72 | |
BC130284.1 | - | 8 - 9 | 1115 | 72 | |
BC130286.1 | - | 8 - 9 | 1115 | 72 | |
M29145.1 | - | 8 - 9 | 1141 | 72 | |
M55379.1 | - | 8 - 9 | 1025 | 72 | |
M60718.1 | - | 8 - 9 | 1112 | 72 | |
M73239.1 | - | 8 - 9 | 1105 | 72 | |
M73240.1 | - | 8 - 9 | 1090 | 72 |