The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242786.1 | NP_001229715.1 | 8 - 9 | 985 | 73 |
NM_001242787.1 | NP_001229716.1 | 8 - 9 | 985 | 73 | |
NM_001242788.1 | NP_001229717.1 | 7 - 8 | 1195 | 73 | |
NM_001242789.1 | NP_001229718.1 | 3 - 4 | 475 | 73 | |
NM_001519.3 | NP_001510.2 | 8 - 9 | 1276 | 73 | |
NM_145685.2 | NP_663718.1 | 4 - 5 | 1184 | 73 | |
XM_005267561.3 | XP_005267618.1 | 8 - 9 | 1298 | 73 | |
XM_005267563.3 | XP_005267620.1 | 4 - 5 | 1988 | 73 | |
XM_006720123.2 | XP_006720186.1 | 3 - 4 | 1416 | 73 | |
XM_011536672.2 | XP_011534974.1 | 8 - 9 | 1298 | 73 | |
XM_011536673.1 | XP_011534975.1 | 7 - 8 | 1148 | 73 | |
XM_011536674.1 | XP_011534976.1 | 4 - 5 | 1566 | 73 | |
GenBank mRNA | AJ297407.1 | - | 4 - 5 | 1192 | 73 |
AK096471.1 | - | 3 - 4 | 475 | 73 | |
AK294899.1 | - | 8 - 9 | 985 | 73 | |
AK295579.1 | - | 8 - 9 | 898 | 73 | |
BC071637.1 | - | 3 - 4 | 340 | 73 | |
BC086856.1 | - | 7 - 8 | 928 | 73 | |
BC103861.2 | - | 4 - 5 | 492 | 73 | |
BC133655.1 | - | 4 - 5 | 492 | 73 | |
U28838.1 | - | 8 - 9 | 1279 | 73 | |
U75276.1 | - | 8 - 9 | 980 | 73 |