Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLA2G12B
Entrez Gene ID
84647
Gene Name
phospholipase A2 group XIIB
Gene Aliases
FKSG71, GXIIB, GXIIIsPLA2, PLA2G13, sPLA2-GXIIB
Chromosome Location
Chr.10: 72934762 - 72955044 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318125.1 NP_001305054.1 2 - 3 512 60
NM_032562.4 NP_115951.2 3 - 4 601 60
XM_005270244.4 XP_005270301.1 3 - 4 814 60
GenBank mRNA AF339053.1 - 3 - 4 560 60
AK311349.1 - 2 - 3 499 60
AK313712.1 - 3 - 4 564 60
AY358032.1 - 3 - 4 534 60
BC093996.1 - 3 - 4 523 60
BC111946.1 - 3 - 4 523 60
DA536331.1 - 2 - 3 499 60

View Full Product Details