The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005120.2 | NP_005111.2 | 31 - 32 | 4616 | 59 |
GenBank mRNA | AB102668.1 | - | 28 - 29 | 3958 | 59 |
AF071309.1 | - | 31 - 32 | 4429 | 59 | |
AK302543.1 | - | 31 - 32 | 4616 | 59 | |
D83783.1 | - | 30 - 31 | 4258 | 59 | |
U80742.1 | - | 5 - 6 | 659 | 59 |