Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MED12
Entrez Gene ID
9968
Gene Name
mediator complex subunit 12
Gene Aliases
ARC240, CAGH45, FGS1, HOPA, MED12S, OHDOX, OKS, OPA1, TNRC11, TRAP230
Chromosome Location
Chr.X: 71118556 - 71142454 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005120.2 NP_005111.2 41 - 42 6242 65
GenBank mRNA AB102668.1 - 38 - 39 5593 65
AF071309.1 - 41 - 42 6052 65
D83783.1 - 40 - 41 5884 65
U80742.1 - 15 - 16 2293 65

View Full Product Details