The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001120.4 | NP_001111.3 | 4 - 5 | 842 | 81 |
NM_001146069.1 | NP_001139541.1 | 5 - 6 | 640 | 81 | |
XM_005247927.3 | XP_005247984.1 | 5 - 6 | 635 | 81 | |
GenBank mRNA | BC001502.2 | - | 5 - 6 | 566 | 81 |
BC014979.1 | - | 5 - 6 | 569 | 81 | |
BX359190.2 | - | 5 - 6 | 641 | 81 | |
L11669.1 | - | 4 - 5 | 648 | 81 |