Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLAMF7
Entrez Gene ID
57823
Gene Name
SLAM family member 7
Gene Aliases
19A, CD319, CRACC, CS1
Chromosome Location
Chr.1: 160739057 - 160754821 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282589.1 NP_001269518.1 3 - 4 717 133
NM_001282590.1 NP_001269519.1 4 - 5 789 133
NM_001282591.1 NP_001269520.1 3 - 4 669 133
NM_001282594.1 NP_001269523.1 4 - 5 828 133
NM_001282595.1 NP_001269524.1 4 - 5 571 133
NM_021181.4 NP_067004.3 5 - 6 1110 133
XM_011509828.1 XP_011508130.1 5 - 6 959 133
GenBank mRNA AB027233.1 - 5 - 6 880 133
AF291815.1 - 5 - 6 876 133
AF390894.1 - 5 - 6 885 133
AJ276429.2 - 5 - 6 898 133
AK292097.1 - 4 - 5 573 133
AK292148.1 - 5 - 6 894 133
AK298499.1 - 3 - 4 524 133
AK301432.1 - 4 - 5 614 133
AK301438.1 - 3 - 4 457 133
AL834424.1 - 4 - 5 578 133
AY358512.1 - 5 - 6 902 133

View Full Product Details