The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003037.3 | NP_003028.1 | 5 - 6 | 1228 | 67 |
XM_005245456.3 | XP_005245513.1 | 4 - 5 | 1364 | 67 | |
XM_011509905.2 | XP_011508207.1 | 5 - 6 | 1449 | 67 | |
XM_017002129.1 | XP_016857618.1 | 6 - 7 | 1537 | 67 | |
XM_017002131.1 | XP_016857620.1 | 6 - 7 | 1532 | 67 | |
GenBank mRNA | AK314244.1 | - | 5 - 6 | 1172 | 67 |
AY040554.1 | - | 5 - 6 | 862 | 67 | |
BC132792.1 | - | 5 - 6 | 894 | 67 | |
KF471075.1 | - | 6 - 7 | 958 | 67 | |
U33017.1 | - | 5 - 6 | 995 | 67 |