The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016540.3 | NP_057624.3 | 3 - 4 | 812 | 67 |
XM_011542559.2 | XP_011540861.1 | 2 - 3 | 746 | 67 | |
GenBank mRNA | AB040973.1 | - | 3 - 4 | 645 | 67 |
AF236081.1 | - | 3 - 4 | 640 | 67 | |
AK315161.1 | - | 3 - 4 | 812 | 67 | |
BC067473.1 | - | 3 - 4 | 640 | 67 | |
BC067474.1 | - | 3 - 4 | 640 | 67 | |
EU432120.1 | - | 3 - 4 | 640 | 67 |