Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TBC1D17
Entrez Gene ID
79735
Gene Name
TBC1 domain family member 17
Gene Aliases
-
Chromosome Location
Chr.19: 49877425 - 49888750 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168222.1 NP_001161694.1 10 - 11 1442 93
NM_024682.2 NP_078958.2 11 - 12 1541 93
XM_011527317.2 XP_011525619.1 11 - 12 1453 93
GenBank mRNA AB449903.1 - 11 - 12 1242 93
AK022230.1 - 11 - 12 1300 93
AK300007.1 - 10 - 11 1201 93
AY129013.1 - 9 - 10 1988 93
BC003516.1 - 11 - 12 1394 93

View Full Product Details