The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_130787.2 | NP_570603.2 | 15 - 16 | 2326 | 105 |
XM_011526557.2 | XP_011524859.1 | 15 - 16 | 2931 | 105 | |
GenBank mRNA | AK302005.1 | - | 12 - 13 | 1640 | 105 |
AL136925.1 | - | 15 - 16 | 2192 | 105 | |
BC014214.2 | - | 15 - 16 | 2224 | 105 |