The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014203.2 | NP_055018.2 | 17 - 18 | 2442 | 68 |
NM_130787.2 | NP_570603.2 | 16 - 17 | 2376 | 68 | |
XM_011526556.2 | XP_011524858.1 | 17 - 18 | 3052 | 68 | |
XM_011526557.2 | XP_011524859.1 | 16 - 17 | 2981 | 68 | |
GenBank mRNA | AK094317.1 | - | 7 - 8 | 878 | 68 |
AL136925.1 | - | 16 - 17 | 2242 | 68 | |
BC014214.2 | - | 16 - 17 | 2274 | 68 |