The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014203.2 | NP_055018.2 | 18 - 19 | 2534 | 73 |
NM_130787.2 | NP_570603.2 | 17 - 18 | 2468 | 73 | |
XM_011526556.2 | XP_011524858.1 | 18 - 19 | 3144 | 73 | |
XM_011526557.2 | XP_011524859.1 | 17 - 18 | 3073 | 73 | |
GenBank mRNA | AK094317.1 | - | 8 - 9 | 970 | 73 |
AL136925.1 | - | 17 - 18 | 2334 | 73 | |
BC014214.2 | - | 17 - 18 | 2366 | 73 |