Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EFHC1
Entrez Gene ID
114327
Gene Name
EF-hand domain containing 1
Gene Aliases
EJM1, dJ304B14.2
Chromosome Location
Chr.6: 52420196 - 52495785 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172420.1 NP_001165891.1 4 - 5 695 146
NM_018100.3 NP_060570.2 3 - 4 783 146
NR_033327.1 3 - 4 783 146
GenBank mRNA AK001328.1 - 3 - 4 669 146
AK225543.1 - 3 - 4 677 146
AK295064.1 - 3 - 4 526 146
AK297632.1 - 4 - 5 695 146
AL122084.1 - 3 - 4 670 146
AY608690.1 - 3 - 4 1237 146
BC020210.1 - 3 - 4 659 146
EU520261.1 - 3 - 4 668 146

View Full Product Details