The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172420.1 | NP_001165891.1 | 6 - 7 | 1044 | 69 |
NM_018100.3 | NP_060570.2 | 5 - 6 | 1132 | 69 | |
NR_033327.1 | 4 - 5 | 2389 | 69 | ||
GenBank mRNA | AK001328.1 | - | 5 - 6 | 1018 | 69 |
AK225543.1 | - | 4 - 5 | 2281 | 69 | |
AK295064.1 | - | 5 - 6 | 875 | 69 | |
AK297632.1 | - | 6 - 7 | 1044 | 69 | |
AL122084.1 | - | 4 - 5 | 2275 | 69 | |
BC020210.1 | - | 5 - 6 | 1008 | 69 | |
EU520261.1 | - | 5 - 6 | 1017 | 69 |