The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172420.1 | NP_001165891.1 | 10 - 11 | 1768 | 54 |
NM_018100.3 | NP_060570.2 | 9 - 10 | 1856 | 54 | |
NR_033327.1 | 8 - 9 | 3113 | 54 | ||
GenBank mRNA | AK001328.1 | - | 9 - 10 | 1742 | 54 |
AK225543.1 | - | 8 - 9 | 3005 | 54 | |
AK295064.1 | - | 9 - 10 | 1599 | 54 | |
AK297632.1 | - | 10 - 11 | 1768 | 54 | |
AL122084.1 | - | 8 - 9 | 2999 | 54 | |
BC020210.1 | - | 9 - 10 | 1732 | 54 | |
EU520261.1 | - | 9 - 10 | 1741 | 54 |