The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144663.1 | NP_001138135.1 | 11 - 12 | 1487 | 145 |
NM_004063.3 | NP_004054.3 | 11 - 12 | 1486 | 145 | |
XM_011516790.1 | XP_011515092.1 | 11 - 12 | 1485 | 145 | |
GenBank mRNA | AK300638.1 | - | 3 - 4 | 365 | 145 |
AK301001.1 | - | 7 - 8 | 812 | 145 | |
AK312873.1 | - | 11 - 12 | 1487 | 145 | |
BC112013.1 | - | 11 - 12 | 1459 | 145 | |
BC113464.1 | - | 11 - 12 | 1447 | 145 | |
U07969.1 | - | 11 - 12 | 1448 | 145 | |
X83228.1 | - | 11 - 12 | 1453 | 145 |