The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144663.1 | NP_001138135.1 | 12 - 13 | 1676 | 97 |
NM_004063.3 | NP_004054.3 | 12 - 13 | 1675 | 97 | |
XM_011516790.1 | XP_011515092.1 | 12 - 13 | 1674 | 97 | |
GenBank mRNA | AK300638.1 | - | 4 - 5 | 554 | 97 |
AK301001.1 | - | 8 - 9 | 1001 | 97 | |
AK312873.1 | - | 12 - 13 | 1676 | 97 | |
BC112013.1 | - | 12 - 13 | 1648 | 97 | |
BC113464.1 | - | 12 - 13 | 1636 | 97 | |
U07969.1 | - | 12 - 13 | 1637 | 97 | |
X83228.1 | - | 12 - 13 | 1642 | 97 |