The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004063.3 | NP_004054.3 | 1 - 2 | 107 | 98 |
XM_011516790.1 | XP_011515092.1 | 1 - 2 | 106 | 98 | |
GenBank mRNA | AK301001.1 | - | 1 - 2 | 75 | 98 |
AU098586.1 | - | 1 - 2 | 120 | 98 | |
BC112013.1 | - | 1 - 2 | 80 | 98 | |
BC113464.1 | - | 1 - 2 | 68 | 98 | |
DB009107.1 | - | 1 - 2 | 106 | 98 | |
U07969.1 | - | 1 - 2 | 69 | 98 | |
X83228.1 | - | 1 - 2 | 74 | 98 |